missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
P2Y12/P2RY12 Polyclonal antibody specifically detects P2Y12/P2RY12 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | P2Y12/P2RY12 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 243-342 of human P2Y12/P2RY12 (NP_795345.1).,, Sequence:, INFLIVIVCYTLITKELYRSYVRTRGVGKVPRKKVNVKVFIIIAVFFICFVPFHFARIPYTLSQTRDVFDCTAENTLFYVKESTLWLTSLNACLDPFIYFFLCKSFRNSLISMLKCPNSATSLSQDNRKKEQDGGDPNEETPM |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?