Learn More
p21/CIP1/CDKN1A Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1047-SP
This item is not returnable.
View return policy
Description
p21/CIP1/CDKN1A Polyclonal specifically detects p21/CIP1/CDKN1A in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Product Title