Learn More
P16-INK4A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10883-1-AP-20UL
 View more versions of this product
Product Code. 16847563
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
10883-1-AP reacts with p16 specifically. P16 plays an essential role in regulating the cell cycle, and mutations in p16 increase the risk of developing various cancers, including melanoma.
Kaiso is a novel member of the BTB/POZ (Broad complex, Tramtrak, Bric a brac/Pox virus and zinc finger) zinc finger superfamily of transcription factors. Yeast 2-hybrid studies have shown that Kaiso interacts with catenin p120ctn. This interaction involves the Arm repeats of 1-7 of the p120ctn Armadillo domain and the carboxyl-terminal third of Kaiso. The significance of the Kaiso:p120 ctn interaction is still unknown.
Specifications
Specifications
P16-INK4A
Polyclonal
Unconjugated
CDKN2A
ARF, ARF,CDKN2A, CDK4 inhibitor p16 INK4, CDK4 inhibitor p16INK4, CDK4I, CDKN2, CDKN2A, CMM2, INK4, INK4a, MLM, MTS 1, MTS1, Multiple tumor suppressor 1, P16, p16 INK4, p16 INK4a, p16INK4, p16INK4a, TP16
Rabbit
Antigen Affinity Chromatography
RUO
1029
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N726
CDKN2A
P16-INK4A Fusion Protein Ag1328