missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Selectin/CD62P Antibody (Psel.KO.2.7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 14 publications
225.00 € - 506.00 €
Specifications
| Antigen | P-Selectin/CD62P |
|---|---|
| Clone | Psel.KO.2.7 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:25-1:50, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18022066
|
Novus Biologicals
NB100-65392SS |
0.025 mg |
225.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18710554
|
Novus Biologicals
NB100-65392 |
0.1 mg |
506.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human, Mouse, Rat, Bovine, Equine, Feline, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| P-Selectin/CD62P | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Bovine, Equine, Feline, Goat, Sheep | |
| CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
| SELP | |
| IgG1 | |
| Protein G purified | |
| NB100-65392 recognizes the P-Selectin cell surface antigen, a 140kD glycoprotein. P-Selectin is expressed by activated platelets and endothelial cells, and plays an important role in adhesive processes between leucocytes and endothelial cells. |
| Psel.KO.2.7 | |
| Flow Cytometry 1:25-1:50, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P16109 | |
| 6403 | |
| P-selectin transfected 300.19 cells. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title