Learn More
p-Nitrophenyl Iodoacetate, TRC
Specifications
Specifications
| Chemical Name or Material | p-Nitrophenyl Iodoacetate |
| CAS | 31252-85-4 |
| Molecular Formula | C8 H6 I N O4 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C8H6INO4/c9-5-8(11)14-7-3-1-6(2-4-7)10(12)13/h1-4H,5H2 |
| SMILES | [O-][N+](=O)c1ccc(OC(=O)CI)cc1 |
| IUPAC Name | (4-nitrophenyl) 2-iodoacetate |
| Molecular Weight (g/mol) | 307.04 |
| Formula Weight | 306.9342 |