Learn More
p-Methyl-cinnamoyl Azide, TRC
Specifications
Specifications
| Chemical Name or Material | p-Methyl-cinnamoyl Azide |
| CAS | 24186-38-7 |
| Molecular Formula | C10H9N3O |
| Quantity | 500 mg |
| Synonym | 3-(4-Methylphenyl)-2-propenoyl Azide |
| InChI Formula | InChI=1S/C10H9N3O/c1-8-2-4-9(5-3-8)6-7-10(14)12-13-11/h2-7H,1H3/b7-6+ |
| SMILES | Cc1ccc(\C=C\C(=O)N=[N+]=[N-])cc1 |
| IUPAC Name | (E)-3-(4-methylphenyl)prop-2-enoyl azide |
| Molecular Weight (g/mol) | 187.2 |
| Formula Weight | 187.0746 |