Learn More
p-Hydroxyphenylacetylglycine, TRC
Specifications
Specifications
| Chemical Name or Material | p-Hydroxyphenylacetylglycine |
| CAS | 28116-23-6 |
| Molecular Formula | C10 H11 N O4 |
| Quantity | 2.5 g |
| Synonym | N-[2-(4-Hydroxyphenyl)acetyl]glycine |
| InChI Formula | InChI=1S/C10H11NO4/c12-8-3-1-7(2-4-8)5-9(13)11-6-10(14)15/h1-4,12H,5-6H2,(H,11,13)(H,14,15) |
| SMILES | OC(=O)CNC(=O)Cc1ccc(O)cc1 |
| IUPAC Name | 2-[[2-(4-hydroxyphenyl)acetyl]amino]acetic acid |
| Molecular Weight (g/mol) | 209.2 |
| Formula Weight | 209.0688 |