Learn More
Melford P-Gal
Product Code. 30018644
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018644 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30018644 Supplier Melford Supplier No. P120005.0
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
  • Substance Name: Phenyl-β-D-galactopyranoside
  • CAS Number: 2818-58-8
  • Molecular Formula: C12H16O6
  • Molecular Weight: 256.26
Specifications
Quantity 5 g