Learn More
p-Ethylphenol, TRC
Product Code. 30369188
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369188 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30369188 Supplier Toronto Research Chemicals Supplier No. TRCE925730250G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Ethylphenol
CAS 123-07-9
Molecular Formula C8 H10 O
Quantity 250 g
Synonym 4-Ethylphenol, Phenol, 4-ethyl- (9CI, ACI), 4-Ethylphenol (ACI), Phenol, p-ethyl- (8CI), (4-Hydroxyphenyl)ethane, (p-Hydroxyphenyl)ethane, 1-Ethyl-4-hydroxybenzene, NSC 62012, p-Ethylphenol, p-Hydroxyethylbenzene
InChI Formula InChI=1S/C8H10O/c1-2-7-3-5-8(9)6-4-7/h3-6,9H,2H2,1H3
SMILES CCc1ccc(O)cc1
IUPAC Name 4-ethylphenol
Molecular Weight (g/mol) 122.16
Formula Weight 122.0732