missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-cadherin Mouse anti-Human, Clone: 1G10F7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66295-1-IG-20UL
This item is not returnable.
View return policy
Description
P-cadherin, also known as Cadherin-3, is a Type 1 cadherin protein that belongs to the cadherin superfamily. Type 1 cadherins are single-pass transmembrane proteins that have 5 extracellular cadherin repeats and an intracellular domain that binds p120-catenin and beta-catenin. Cadherins are calcium-dependent cell-cell adhesion glycoproteins responsible for a range of processes including development, wound healing, cell-cell signaling, cell growth and differentiation. P-cadherin is expressed in human fetal structures, and in adult tissues such as the basal layer of the epidermis, breast, prostate, ovary, cervix, hair follicle, and corneal endothelium. P-cadherin expression has also been reported on embryonic stem cells, and stem cells of the normal mammary gland, and hair follicle. Overexpression of P-cadherin has been associated with poor prognosis in breast, prostate, ovary, colon, and stomach carcinomas. Mutations in CDH3 are associated with congential hypotrichosis with juvenile macular dystrophy.Specifications
| P-cadherin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22223 | |
| CDH3 | |
| P-cadherin Fusion Protein Ag4758 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1G10F7 | |
| Unconjugated | |
| CDH3 | |
| Cadherin 3, CDH3, CDHP, HJMD, P cadherin, PCAD, P-cadherin, Placental cadherin | |
| Mouse | |
| Protein G | |
| RUO | |
| 1001 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction