Learn More
p-Benzyloxybenzyl Alcohol, TRC
Specifications
Specifications
| Chemical Name or Material | p-Benzyloxybenzyl Alcohol |
| CAS | 836-43-1 |
| Molecular Formula | C14H14O2 |
| Quantity | 25 g |
| Synonym | (4-Benzyloxyphenyl)methanol, 4-(Benzyloxy)benzyl Alcohol, 4-(Phenylmethoxy)benzenemethanol, NSC 131675 |
| InChI Formula | InChI=1S/C14H14O2/c15-10-12-6-8-14(9-7-12)16-11-13-4-2-1-3-5-13/h1-9,15H,10-11H2 |
| SMILES | OCc1ccc(OCc2ccccc2)cc1 |
| IUPAC Name | (4-phenylmethoxyphenyl)methanol |
| Molecular Weight (g/mol) | 214.26 |
| Formula Weight | 214.0994 |