Learn More
p-(Allyloxy)benzyl Alcohol, TRC
Specifications
Specifications
| Chemical Name or Material | p-(Allyloxy)benzyl Alcohol |
| CAS | 3256-45-9 |
| Molecular Formula | C10H12O2 |
| Quantity | 100 mg |
| Synonym | 4-(2-Propenyloxy)benzenemethanol, 4-(Allyloxy)benzyl Alcohol |
| InChI Formula | InChI=1S/C10H12O2/c1-2-7-12-10-5-3-9(8-11)4-6-10/h2-6,11H,1,7-8H2 |
| SMILES | C=CCOC1=CC=C(CO)C=C1 |
| IUPAC Name | (4-prop-2-enoxyphenyl)methanol |
| Molecular Weight (g/mol) | 164.2 |
| Formula Weight | 164.08 |