Learn More
Ozenoxacin, >90%, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Ozenoxacin |
| CAS | 245765-41-7 |
| Molecular Formula | C21 H21 N3 O3 |
| Quantity | 25 mg |
| Synonym | 3-Quinolinecarboxylic acid','1-cyclopropyl-1,4-dihydro-8-methyl-7-[5-methyl-6-(methylamino)-3-pyridinyl]-4-oxo-, 1-Cyclopropyl-1,4-dihydro-8-methyl-7-[5-methyl-6-(methylamino)-3-pyridinyl]-4-oxo-3-quinolinecarboxylic acid, 1-Cyclopropyl-8-methyl-7-[5-methyl-6-(methylamino)-3-pyridinyl]-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid, Ozenoxacin, T 3912, Xepi |
| InChI Formula | InChI=1S/C21H21N3O3/c1-11-8-13(9-23-20(11)22-3)15-6-7-16-18(12(15)2)24(14-4-5-14)10-17(19(16)25)21(26)27/h6-10,14H,4-5H2,1-3H3,(H,22,23)(H,26,27) |
| SMILES | CNc1ncc(cc1C)c2ccc3C(=O)C(=CN(C4CC4)c3c2C)C(=O)O |
| IUPAC Name | 1-cyclopropyl-8-methyl-7-[5-methyl-6-(methylamino)pyridin-3-yl]-4-oxoquinoline-3-carboxylic acid |
| Molecular Weight (g/mol) | 363.4097 |
| Formula Weight | 363.1583 |