Learn More
Oxyisofenphos, TRC
Specifications
Specifications
| Chemical Name or Material | Isofenphos-oxon |
| CAS | 31120-85-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 H24 N O5 P |
| Quantity | 1 g |
| Synonym | Salicylic acid','isopropyl ester','ethyl isopropylphosphoramidate (8CI), Isofenphos oxon, Oxyisofenphos, Benzoic acid','2-[[ethoxy[(1-methylethyl)amino]phosphinyl]oxy]-','1-methylethyl ester |
| InChI Formula | InChI=1S/C15H24NO5P/c1-6-19-22(18,16-11(2)3)21-14-10-8-7-9-13(14)15(17)20-12(4)5/h7-12H,6H2,1-5H3,(H,16,18) |
| SMILES | CCOP(=O)(NC(C)C)Oc1ccccc1C(=O)OC(C)C |
| IUPAC Name | propan-2-yl 2-[ethoxy-(propan-2-ylamino)phosphoryl]oxybenzoate |
| Molecular Weight (g/mol) | 329.33 |
| Show More |