Learn More
Oxolinic Acid-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Oxolinic acid ethyl-d5 |
| CAS | 1189890-98-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13 2H5 H6 N O5 |
| Quantity | 10 mg |
| Synonym | 8-Oxo-5-(1,1,2,2,2-pentadeuterioethyl)-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid |
| InChI Formula | InChI=1S/C13H11NO5/c1-2-14-5-8(13(16)17)12(15)7-3-10-11(4-9(7)14)19-6-18-10/h3-5H,2,6H2,1H3,(H,16,17)/i1D3,2D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])N1C=C(C(=O)O)C(=O)c2cc3OCOc3cc12 |
| IUPAC Name | 8-oxo-5-(1,1,2,2,2-pentadeuterioethyl)-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid |
| Molecular Weight (g/mol) | 266.26 |
| Show More |