Learn More
Thermo Scientific™ Oxoid™ PALCAM Selective Supplement
Description
Oxoid PALCAM Selective Supplement is used for the detection of Listeria monocytogenes.
Add to PALCAM Agar Base, Part No. CM0877B
Specifications
Specifications
| Description | PALCAM Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |