missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ Oxivir™ Plus NC Spray

Product Code. 17976710
Change view
Click to view available options
Volume (Metric):
0.75 L
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17976710 0.75 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17976710 Supplier Diversey™ Supplier No. 101106359
  • ONLINE EXCLUSIVE PRICE
    98.07 €
    88.62 € / Pack of 6
    Save 9.45 €
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxivir™ Plus NC Spray is a broad spectrum cleaner and disinfectant for both food and non-food, non-porous, hard surfaces.

  • Bactericidal, Virucidal, Yeasticidal, Fungicidal
  • Patented Hydrogen Peroxide based acidic formulation
  • Short contact times
  • Effective cleaner
  • Fragrance free
  • Suitable for cleaning and disinfecting of various water resistant surfaces in food premises in all water conditions
  • Extensively tested against European norms and effective against multiple pathogens of concern, including all enveloped and non-enveloped viruses such as Coronavirus or Norovirus, but also bacterias, years and fungi
  • High cleaning performance, including descaling and lime limitation removes the need for a pre-cleaning step
  • Good user safety profile, with no hazard warnings on RTU format
  • Does not contain respiratory irritants
  • AHP formulation is gentle on surfaces extending equipment life
  • Good environmental profile
  • Actively breaks down into oxygen and water
  • For use in multiple sectors including schools, universities, hotels, holiday parks, gyms, bars, offices, receptions, staff, areas, dining rooms, bars, pubs, restaurants, shops, malls

Specifications

Product Type Disinfectant
Color-coded No
Form Liquid
pH Neutral
Product Line Oxivir
Scent Unscented
Sterility Non-sterile
Appearance Clear
Color Clear
Disposable Single-use
Filtered Non-filtered
Flammable No
Quantity 6 Bottles
Volume (Metric) 0.75 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.