missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ Oxivir Excel Concentrate

Product Code. 17241614
Change view
Click to view available options
Description:
For AT only
For AT, CH, DE, DK, FI, FR, GB, IE, NO, SE only
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
17241614 For AT only Pack of 2
17231614 For AT, CH, DE, DK, FI, FR, GB, IE, NO, SE only Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17241614 Supplier Diversey™ Supplier No. 100968158
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly concentrated one step cleaner disinfectant for non-invasive medical devices and nonporous surfaces.

  • Effective against Bacteria, Viruses and Yeasts
  • Full virucidal claim including non-enveloped and enveloped viruses such as SARS-CoV-2 in just 30 sec
  • Extensively tested against European norms
  • Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
  • Excellent user safety profile reduces worker risk
  • No hazard warnings in dilution
  • Based on Diversey's patented AHP formulation (Accelerated Hydrogen Peroxide)
  • AHP formulation is gentle on surfaces extending equipment life
  • Does not contain respiratory irritants
  • Medical Device Class IIa
  • Oxivir Excel can be used on any hard surface usually encountered in a laboratory, production and non-production environments including stainless steel, glass, acrylics and PVC. It is not suitable for use on marble, copper, brass or other surfaces sensitive to oxidative agents.

Spécification

Product Type Disinfectant Concentrate
pH 1.95
Sterility Non-sterile
Composition Patented Accelerated Hydrogen Peroxide formulation (conc.)
Container Type Jug
Description For AT only
Quantity 2 Pk.
Volume (Metric) 5 L

The surfactants used in this product are biodegradable in compliance with the requirement of EU Detergent Regulation, EC 648/2004

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.