Learn More
Oxindole-3-acetic Acid-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | Oxindole-3-acetic Acid-13C6 |
| CAS | 2724918-56-1 |
| Molecular Formula | 13C6 C4 H9 N O3 |
| Quantity | 1 mg |
| Synonym | 2-Oxo-3-Indolineacetic-13C6 Acid, (±)-2,3-Dihydro-2-oxoindole-3-acetic-13C6 Acid, (±)-2-Oxindoline-3-acetic-13C6 Acid, (±)-2-Oxo-3-indolineacetic-13C6 Acid, 2-(2-Oxoindolin-3-yl)acetic-13C6 Acid |
| InChI Formula | InChI=1S/C10H9NO3/c12-9(13)5-7-6-3-1-2-4-8(6)11-10(7)14/h1-4,7H,5H2,(H,11,14)(H,12,13)/i1+1,2+1,3+1,4+1,6+1,8+1 |
| SMILES | C(C(O)=O)C1[13C]=2[13C](NC1=O)=[13CH][13CH]=[13CH][13CH]2 |
| IUPAC Name | 2-(2-oxo-1,3-dihydroindol-3-yl)acetic acid |
| Molecular Weight (g/mol) | 197.139 |
| Formula Weight | 197.078 |