missing translation for 'onlineSavingsMsg'
Learn More

Oxidopamine hydrobromide, MedChemExpress

Product Code. 16402256
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16402256

Brand: Medchem Express HYB1081A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxidopamine (6-OHDA) hydrobromide is an antagonist of the neurotransmitter dopamine.

Oxidopamine hydrobromide is a widely used neurotoxin and selectively destroys dopaminergic neurons. Oxidopamine hydrobromide promotes COX-2 activation, leading to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion. Oxidopamine hydrobromide can be used for the research of Parkinson’s disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome.

Specifications

Chemical Name or Material Oxidopamine hydrobromide
CAS 636-00-0
Color Gray
Purity Grade Notes Research
Molecular Formula C8H12BrNO3
Quantity 500 mg
Synonym 6-Hydroxydopamine hydrobromide 6-OHDA hydrobromide
Solubility Information DMSO : 50 mg/mL (199.93 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 20 mg/mL (79.97 mM; Need ultrasonic)
SMILES OC1=CC(CCN)=C(O)C=C1O.[H]Br
Molecular Weight (g/mol) 250.09
Formula Weight 250.09
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.