Learn More
Oxiconazole nitrate, MedChemExpress
Product Code. 16498726
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16498726 100 mg 100mg
16414587 50 mg 50mg
16408698 10 mg 10mg
3 options
Product Code. 16498726
Brand: Medchem Express HYB1324
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Oxiconazole (Ro 13-8996) nitrate is a broad spectrum anti-fungal agent which can inhibit the growth of Candida, Aspergillus and Trichophyton.
Oxiconazole nitrate is also a highly efficacious activator of CYP3A4 transactivation, which could be antagonized by Rifampicin (HY-B0272) in a competitive manner. Oxiconazole nitrate exhibits inhibitory effect against colorectal cancer (CRC) via peroxiredoxin-2 (PRDX2)-mediated autophagy arrest.
Specifications
Chemical Name or Material Oxiconazole nitrate
Physical Form Solid
CAS 64211-46-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H14Cl4N4O4
For Use With (Application) COVID-19-immunoregulation
Synonym Ro 13-8996
Solubility Information DMSO : ≥ 100 mg/mL (203.19 mM)
SMILES [O-][N+](O)=O.ClC1=CC=C(/C(CN2C=CN=C2)=N/OCC3=CC=C(Cl)C=C3Cl)C(Cl)=C1
Molecular Weight (g/mol) 492.14
Formula Weight 492.14
Percent Purity 98.0%
Grade Research