Learn More
Tocris Bioscience™ OXF BD 02
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
OXF BD 02 is a selective inhibitor of the first bromodomain of BRD4 (BRD4(1)) (IC50 = 382 nM). Exhibits 2-3-fold selectivity for BRD4(1) over the CBP bromodomain and has little affinity for a range of other bromodomains. Reduces viability of lung adenocarcinoma cell lines and attenuates proliferation of MV-4-11 leukemia cells. Cell permeable.
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | OXF BD 02 |
| Molecular Weight (g/mol) | 295.33 |
| Inhibitors | OXF BD 02 |