Learn More
Tocris Bioscience™ OXA 06 dihydrochloride
Potent ROCK inhibitor
Brand: Tocris Bioscience™ 5182/10
This item is not returnable.
View return policy
Description
OXA 06 dihydrochloride is a potent ROCK inhibitor (IC50 = 10 nM). Suppresses pMYPT1 and pCofilin levels in non-small cell lung carcinoma (NSCLC) cell lines. Also inhibits anchorage-independent growth of NSCLC cell lines in vitro.Specifications
| >98% | |
| 1825455-91-1 | |
| 10 mg |