Learn More
Karl Hecht™ Overflow Volumetric Flask
Click to view available options
Capacity (Metric):
22.7 mL
43.5 mL
97 mL
164 mL
250 mL
365 mL
432 mL
Unit Size:
Each
Description
- Thick-walled, solid finish.
- Clearly legible figures and marks.
Specifications
Specifications
| Capacity (Metric) | 164 mL |
| Material | Borosilicate Glass |
| Product Type | Volumetric Flask |