Learn More
OSW-1 (Orsaponin), TRC
Specifications
Specifications
| Chemical Name or Material | Orsaponin |
| CAS | 145075-81-6 |
| Molecular Formula | C47H68O15 |
| Quantity | 2.5 mg |
| Synonym | (3β,16β)-16-[[2-O-Acetyl-3-O-[2-O-(4-methoxybenzoyl)-β-D-xylopyranosyl]-α-L-arabinopyranosyl]oxy]-3,17-dihydroxycholest-5-en-22-one, Orsaponin |
| InChI Formula | InChI=1S/C47H68O15/c1-24(2)8-15-34(50)25(3)47(55)37(21-33-31-14-11-28-20-29(49)16-18-45(28,5)32(31)17-19-46(33,47)6)60-44-41(59-26(4)48)39(36(52)23-58-44)62-43-40(38(53)35(51)22-57-43)61-42(54)27-9-12-30(56-7)13-10-27/h9-13,24-25,29,31-33,35-41,43-44,49,51-53,55H,8,14-23H2,1-7H3/t25-,29+,31-,32+,33+,35-,36+,37+,38+,39+,40-,41-,43+,44+,45+,46+,47-/m1/s1 |
| SMILES | C[C@@]12[C@](C[C@@H]([C@]2(O)[C@H](C)C(CCC(C)C)=O)O[C@H]3[C@@H]([C@H]([C@H](CO3)O)O[C@H]4[C@@H]([C@H]([C@@H](CO4)O)O)OC(C5=CC=C(C=C5)OC)=O)OC(C)=O)([H])[C@@]6([H])[C@]([C@@]7(C(C[C@H](CC7)O)=CC6)C)([H])CC1 |
| IUPAC Name | (2S,3R,4S,5R)-2-(((2S,3R,4S,5S)-3-acetoxy-2-(((3S,8R,9S,10R,13S,14S,16S,17S)-3,17-dihydroxy-10,13-dimethyl-17-((S)-6-methyl-3-oxoheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-16-yl)oxy)-5-hydroxytetrahydro-2H-pyran-4-yl)oxy)-4,5-dihydroxytetrahydro-2H-pyran-3-yl 4-methoxybenzoate |
| Molecular Weight (g/mol) | 873.03 |
| Formula Weight | 872.46 |