missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSU-T315, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
OSU-T315 (ILK-IN-1) is a small Integrin-linked kinase (ILK) inhibitor with an IC50 of 0.6 μM, inhibiting PI3K/AKT signaling by dephosphorylation of AKT-Ser473 and other ILK targets (GSK-3β and myosin light chain). OSU-T315 abrogates AKT activation by impeding AKT localization in lipid rafts and triggers caspase-dependent apoptosis in an ILK-independent manner. OSU-T315 causes cell death through apoptosis and autophagy.
Specifications
Specifications
| Chemical Name or Material | OSU-T315 |
| Physical Form | Solid |
| CAS | 2070015-22-2 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C30H30F3N5O |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : ≥ 260 mg/mL (487.27 mM) |
| SMILES | O=C(NC)CCC1=CC(C2=CC=C(C3=CC=C(C(F)(F)F)C=C3)C=C2)=NN1C4=CC=C(N5CCNCC5)C=C4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction