missing translation for 'onlineSavingsMsg'
Learn More

OSS_128167, MedChemExpress

Product Code. 16444138
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444138

Brand: Medchem Express HY107454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OSS_128167 is a potent selective sirtuin 6 (SIRT6) inhibitor with IC50s of 89 μM, 1578 μM and 751 μM for SIRT6, SIRT1 and SIRT2, respectively. OSS_128167 has anti-HBV activity that inhibits HBV transcription and replication. OSS_128167 has anti-acncer, anti-inflammation and anti-viral effects.

OSS_128167 is a potent selective sirtuin 6 (SIRT6) inhibitor with IC50s of 89 μM, 1578 μM and 751 μM for SIRT6, SIRT1 and SIRT2, respectively. OSS_128167 has anti-HBV activity that inhibits HBV transcription and replication. OSS_128167 has anti-acncer, anti-inflammation and anti-viral effects.

Specifications

Chemical Name or Material OSS_128167
Physical Form Solid
CAS 887686-02-4
Color Tan
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H14N2O6
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 103.3 mg/mL (281.99 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC(NC(C2=CC=CO2)=O)=CC=C1)NC3=CC(C(O)=O)=C(O)C=C3
Molecular Weight (g/mol) 366.32
Formula Weight 366.32
Percent Purity 98.06%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.