Learn More
OSS_128167, MedChemExpress
Product Code. 16480487
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16480487
Brand: Medchem Express HY107454
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress OSS_128167 is a potent selective sirtuin 6 (SIRT6) inhibitor with IC50s of 89 μM, 1578 μM and 751 μM for SIRT6, SIRT1 and SIRT2, respectively. OSS_128167 has anti-HBV activity that inhibits HBV transcription and replication. OSS_128167 has anti-acncer, anti-inflammation and anti-viral effects.
OSS_128167 is a potent selective sirtuin 6 (SIRT6) inhibitor with IC50s of 89 μM, 1578 μM and 751 μM for SIRT6, SIRT1 and SIRT2, respectively. OSS_128167 has anti-HBV activity that inhibits HBV transcription and replication. OSS_128167 has anti-acncer, anti-inflammation and anti-viral effects.
Specifications
Chemical Name or Material OSS_128167
Physical Form Solid
CAS 887686-02-4
Color Tan
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H14N2O6
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 103.3 mg/mL (281.99 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC(NC(C2=CC=CO2)=O)=CC=C1)NC3=CC(C(O)=O)=C(O)C=C3
Molecular Weight (g/mol) 366.32
Formula Weight 366.32
Percent Purity 98.06%
Grade Research