missing translation for 'onlineSavingsMsg'
Learn More

OSR2 Antibody, Novus Biologicals™

Product Code. 18151174 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18151174 0.1 mL 0.1mL
18476781 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18151174 Supplier Novus Biologicals Supplier No. NBP230669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OSR2 Polyclonal specifically detects OSR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSR2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunodiffusion
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunodiffusion
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q8N2R0
Gene Alias odd-skipped related 2 (Drosophila), protein odd-skipped-related 2
Gene Symbols OSR2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: TMHMNHWTLGYPNVHEITRSTITEMAAAQGLVDARFPFPALPFTTHLFHPKQGAIAHVLPALHKDRPRFDFANL
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 116039
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.