missing translation for 'onlineSavingsMsg'
Learn More

Osmium Tetroxide (4% in H2O), TRC

Product Code. 30133698
Change view
Click to view available options
Quantity:
1 mL
2.5 mL
Unit Size:
1mL
2.5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133698 1 mL 1mL
30011766 2.5 mL 2.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133698 Supplier Toronto Research Chemicals Supplier No. TRCO7025051ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Formula Weight 255.9411
InChI Formula InChI=1S/4O.Os
IUPAC Name tetraoxoosmium
Molecular Formula O4 Os
Molecular Weight (g/mol) 254.23
SMILES O=[Os](=O)(=O)=O
Synonym Osmium oxide (OsO4), (T-4)-, Osmium oxide (OsO4) (8CI), Milas' reagent, Osmium oxide, Osmium tetraoxide, Osmium tetroxide, Osmium(IV) oxide, Reagents, Milas', Tetraoxoosmium
Concentration or Composition (by Analyte or Components) Osmium Tetroxide 4 %
Chemical Name or Material Osmium Tetroxide

Specifications

Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.