Learn More
OSIP-486823, MedChemExpress
Product Code. 16422298
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 16422298
Brand: Medchem Express HYU00004
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress OSIP-486823 is a novel microtubule-interfering agent with distinct biological effects on both protein kinase G (PKG) and microtubules.
OSIP-486823 is a novel microtubule-interfering agent with distinct biological effects on both protein kinase G (PKG) and microtubules.
Specifications
Chemical Name or Material OSIP-486823
CAS 200803-37-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H28FNO4
Quantity 5 mg
Synonym OSIP 486823OSIP486823CP248
SMILES O=C(NCC1=CC=CC=C1)CC(C2=C/3C=CC(F)=C2)=C(C)C3=C\C4=CC(OC)=C(OC)C(OC)=C4
Molecular Weight (g/mol) 473.54
Formula Weight 473.54
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease