missing translation for 'onlineSavingsMsg'
Learn More

OSI-930, MedChemExpress

Product Code. 16430087
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16430087

Brand: Medchem Express HY10204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OSI-930 is an orally selective inhibitor of Kit, KDR and CSF-1R (c-Fms) with IC50s of 80 nM, 9 nM and 15 nM, respectively. OSI-930 also moderately inhibits Flt-1, c-Raf, Lck and low activity against PDGFRα/β, Flt-3 and Abl. OSI-930 has antitumor activity.

OSI-930 is an orally selective inhibitor of Kit, KDR and CSF-1R (c-Fms) with IC50s of 80 nM, 9 nM and 15 nM, respectively. OSI-930 also moderately inhibits Flt-1, c-Raf, Lck and low activity against PDGFRα/β, Flt-3 and Abl. OSI-930 has antitumor activity.

Specifications

Chemical Name or Material OSI-930
CAS 728033-96-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H16F3N3O2S
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (112.75 mM; Need ultrasonic)
SMILES O=C(C1=C(NCC2=CC=NC3=CC=CC=C23)C=CS1)NC4=CC=C(OC(F)(F)F)C=C4
Molecular Weight (g/mol) 443.44
Formula Weight 443.44
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.