Learn More
Orteronel, MedChemExpress
Product Code. 16470367
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16470367
Brand: Medchem Express HY10505
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Orteronel (TAK-700) is a highly selective inhibitor of human 17,20-lyase (CYP17) with IC50 of 38 nM, and exhibits >1000-fold selectivity over other CYPs such as 11-hydroxylase and CYP3A4.
Orteronel (TAK-700) is a highly selective inhibitor of human 17,20-lyase (CYP17) with IC50 of 38 nM, and exhibits >1000-fold selectivity over other CYPs such as 11-hydroxylase and CYP3A4.
Specifications
Chemical Name or Material Orteronel
CAS 566939-85-3
Color White
Purity Grade Notes Research
Molecular Formula C18H17N3O2
Quantity 50 mg
Synonym TAK-700
Solubility Information DMSO : 14.29 mg/mL (46.49 mM; Need ultrasonic)
SMILES O=C(C1=CC=C2C=C([C@@]3(O)CCN4C=NC=C43)C=CC2=C1)NC
Molecular Weight (g/mol) 307.35
Formula Weight 307.35
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death