missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP150/HSP12A Antibody (6G7-2H5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59346-25ug
This item is not returnable.
View return policy
Description
ORP150/HSP12A Monoclonal specifically detects ORP150/HSP12A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ORP150/HSP12A | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.1% Sodium Azide | |
| HYOU1 | |
| Raised against a synthetic peptide of human GRP170 | |
| 25 μg | |
| Hypoxia | |
| 10525 | |
| Store at -20°C. | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 6G7-2H5 | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| 150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects 170kDa. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction