missing translation for 'onlineSavingsMsg'
Learn More

Soblidotin, MedChemExpress

Product Code. 16421439
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16421439 1 mg 1mg
16476128 10 mg 10mg
16453436 25 mg 25mg
16452267 50 mg 50mg
16408737 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16421439 Supplier Medchem Express Supplier No. HY14672

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Soblidotin (Auristatin PE) is a novel synthetic Dolastatin 10 derivative and inhibitor of tubulin polymerization.

Soblidotin (Auristatin PE) is a novel synthetic Dolastatin 10 derivative and inhibitor of tubulin polymerization.

Specifications

Chemical Name or Material Soblidotin
Physical Form Solid
CAS 149606-27-9
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C39H67N5O6
For Use With (Application) Cancer-Kinase/protease
Synonym Auristatin PE TZT-1027
Solubility Information DMSO : 100 mg/mL (142.45 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N[C@@H](C(C)C)C(N([C@@H]([C@H](CC)C)[C@H](OC)CC(N1CCC[C@@]1([H])[C@H](OC)[C@@H](C)C(NCCC2=CC=CC=C2)=O)=O)C)=O)[C@H](C(C)C)N(C)C
Molecular Weight (g/mol) 701.98
Formula Weight 701.98
Percent Purity 99.39%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.