Learn More
Orotidine, TRC
Product Code. 30006994
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006994 5 mg 5mg
30003938 10 mg 10mg
30005778 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006994 Supplier Toronto Research Chemicals Supplier No. TRCO6915205MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 314-50-1
Molecular Formula C10 H12 N2 O8
Molecular Weight (g/mol) 288.21
Synonym 4-Pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-3-β-D-ribofuranosyl-, Orotidine (6CI,7CI,8CI), 1,2,3,6-Tetrahydro-2,6-dioxo-3-β-D-ribofuranosyl-4-pyrimidinecarboxylic acid, 6-Carboxyuridine, Orotic acid riboside
IUPAC Name 3-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2,6-dioxopyrimidine-4-carboxylic acid
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C(=O)NC(=O)C=C2C(=O)O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C10H12N2O8/c13-2-4-6(15)7(16)8(20-4)12-3(9(17)18)1-5(14)11-10(12)19/h1,4,6-8,13,15-16H,2H2,(H,17,18)(H,11,14,19)/t4-,6-,7-,8-/m1/s1
Formula Weight 288.0594
Percent Purity >95%
Chemical Name or Material Orotidine
Grade HPLC