Learn More
Invitrogen™ ORMDL3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA590144
Product Code. 16597691
  • 520.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Immunogen sequence: MNVGTAHSEV NPNTRVMNSR GIWLSYVLAI GLLHIVLLSI PFVSVPVVWT LTNLIHNMGM YIFLHTVKGT PFETPDQGKA RLLTHWEQMD YGVQFTASRK; Positive Samples: 293T, Mouse eye, Mouse thymus, Mouse liver; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.
Negative regulator of sphingolipid synthesis. May indirectly regulate endoplasmic reticulum-mediated Ca(+2) signaling.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ORMDL3
Polyclonal
Unconjugated
ORMDL3
2810011N17Rik; AI315223; ORM1-like 3 (S. cerevisiae); ORM1-like protein 3; ORMDL sphingolipid biosynthesis regulator 3; Ormdl3
Rabbit
Affinity Chromatography
RUO
66612, 94103
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
2.22 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q8N138, Q9CPZ6
ORMDL3
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ORMDL3 (NP_6448091).
100 μL
Primary
Human, Mouse