Learn More
Invitrogen™ ORMDL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590144
Description
Immunogen sequence: MNVGTAHSEV NPNTRVMNSR GIWLSYVLAI GLLHIVLLSI PFVSVPVVWT LTNLIHNMGM YIFLHTVKGT PFETPDQGKA RLLTHWEQMD YGVQFTASRK; Positive Samples: 293T, Mouse eye, Mouse thymus, Mouse liver; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.
Negative regulator of sphingolipid synthesis. May indirectly regulate endoplasmic reticulum-mediated Ca(+2) signaling.
Specifications
| ORMDL3 | |
| Polyclonal | |
| Unconjugated | |
| ORMDL3 | |
| 2810011N17Rik; AI315223; ORM1-like 3 (S. cerevisiae); ORM1-like protein 3; ORMDL sphingolipid biosynthesis regulator 3; Ormdl3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 66612, 94103 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.22 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8N138, Q9CPZ6 | |
| ORMDL3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ORMDL3 (NP_6448091). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.