missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORL1/OPRL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93327-0.1ml
This item is not returnable.
View return policy
Description
ORL1/OPRL1 Polyclonal antibody specifically detects ORL1/OPRL1 in Human samples. It is validated for Western Blot
Specifications
| ORL1/OPRL1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Kappa-type 3 opioid receptor, KOR-3MGC34578, nociceptin receptor, NOCIR, OORORL1kappa3-related opioid receptor, opiate receptor-like 1, Orphanin FQ receptor | |
| A synthetic peptide corresponding to a sequence within amino acids 270-370 of human ORL1/OPRL1 (NP_000904.1). AVFVGCWTPVQVFVLAQGLGVQPSSETAVAILRFCTALGYVNSCLNPILYAFLDENFKACFRKFCCASALRRDVQVSDRVRSIAKDVALACKTSETVPRPA | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 4987 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction