Learn More
Thermo Scientific™ Orion™ BNC Meter Shorting Strap
Click to view available options
:
BNC Shorting Cap
Description
Use the Thermo Scientific™ Orion™ BNC Meter Shorting Strap on pH meters with BNC input to perform BNC input verification testing and meter self test.
Specifications
Specifications
| Description | Orion BNC shorting cap for pH, ORP and ISE meters |
| Type | Meter Accessories |
| Includes | One BNC shorting cap |