missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer Accessories and Kits

Product Code. 13147583 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Electrode Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
13147583 Electrode Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13147583 Supplier Thermo Scientific™ Supplier No. 2120XPEK
  • ONLINE EXCLUSIVE PRICE
    1395.00 €
    1351.34 € / 1 sets
    Save 43.66 €
    3% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accessories and kits for the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer include a consumables kit, calibration kit, carrying case, electrode kit and faceplate.

Use the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer Accessories and Kits with the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer. Accessories and kits include a calcium consumables kit, a reagent-less calcium hardness kit, a calcium hardness calibration kit with carrying case, a calcium electrode kit or a electronics faceplate.

TRUSTED_SUSTAINABILITY

Specifications

Description Calcium Electrode Kit - includes calcium electrode, reference electrode and 2 oz. reference electrode filling solution
Temperature (Metric) Operating 5°C to 45°C (Ambient)
Type Electrode Kit
For Use With (Equipment) Orion™ 2120XP Calcium Hardness Analyzer
Includes Calcium electrode, reference electrode and 2 oz. reference electrode filling solution

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.