Learn More
B Braun™ Original Perfusor™ Syringes with Aspiration Needle
Product Code. 10294072
Click to view available options
Volume (Metric):
20 mL
50 mL
Unit Size:
Each
Pack of 100
This item is not returnable. View return policy
Product Code. 10294072
Brand: B Braun™ 8728810F
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Use this 3-piece syringe for infusion pumps. B Braun™ Original Perfusor™ Syringes with Aspiration enable slow patient during infusion of drug solutions.
  • Polypropylene
  • High contrast graduation
  • Prmanent markings for readability
  • Graduation in ml increments
Specifications
Product Type Original Perfusor Syringe
Quantity 100 Pk.
Volume (Metric) 50 mL