Learn More
B Braun™ Original Perfusor™ Syringes
Click to view available options
Unit Size:
Pack of 100
Description
3-piece syringe for infusion pumps
- Made of polypropylene
- Highly transparent barrel
- High contrast graduation (permanent markings, ideal readability, graduation in ml)
- Centric Luer Lock cone for the attachment of a needle or for connection with other medical products (infusion tubing)
- Oval grip plate for better handling of the syringe and for prevention of rolling
- Safe plunger backstop
- Minimal residual volume
- Plunger stopper with double sealing ring for slow aspiration or infusion of drug solutions, made of synthetic material (natural latex-free)
- Available with aspiration needle
Specifications
Specifications
| Material | Polypropylene |
| For Use With (Application) | Infusion pumps |
| Includes | Without aspiration needle |
| Quantity | 100/Pk. |
| Packaging | 100 Pack |
| Volume (Metric) | 50 mL |
| Connection Type | Luer Lock |
| Graduated | Yes |