missing translation for 'onlineSavingsMsg'
Learn More

Orientin, TRC

Product Code. 30009261
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009261 100 mg 100mg
30133475 50 mg 50mg
30010856 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009261 Supplier Toronto Research Chemicals Supplier No. TRCO674900100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28608-75-5
Molecular Formula C21 H20 O11
Molecular Weight (g/mol) 448.38
Synonym Orientin (6CI,7CI,8CI), 4H-1-Benzopyran-4-one, 2-(3,4-dihydroxyphenyl)-8-β-D-glucopyranosyl-5,7-dihydroxy-, 2-(3,4-Dihydroxyphenyl)-8-β-D-glucopyranosyl-5,7-dihydroxy-4H-1-benzopyran-4-one, 8-β-D-Glucopyranosyl-3',4',5,7-tetrahydroxyflavone, Luteolin 8-C-glucoside, Luteolin 8-C-β-D-glucopyranoside, Luteolin 8-C-β-glucopyranoside, Luteolin-8-C-glucoside, Luteolin-8-glucoside, Lutexin, Orientin (flavone)
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]chromen-4-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc(O)c3C(=O)C=C(Oc23)c4ccc(O)c(O)c4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H20O11/c22-6-14-17(28)18(29)19(30)21(32-14)16-11(26)4-10(25)15-12(27)5-13(31-20(15)16)7-1-2-8(23)9(24)3-7/h1-5,14,17-19,21-26,28-30H,6H2/t14-,17-,18+,19-,21+/m1/s1
Formula Weight 448.1006
Percent Purity >95%
Chemical Name or Material Orientin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.