missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orexin B/Hypocretin-2 Antibody (MM0501-5G21), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11834AF750
This item is not returnable.
View return policy
Description
Orexin B/Hypocretin-2 Monoclonal specifically detects Orexin B/Hypocretin-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Orexin B/Hypocretin-2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HCRT | |
| Human Orexin B/Hypocretin-2 peptide | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MM0501-5G21 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| HCRT, Hcrt 2, Hcrt2, Hypocretin, Hypocretin (orexin) neuropeptide precursor, Hypocretin 2, Hypocretin B, Hypocretin precursor, Hypocretin2, Orexin, Orexin neuropeptide precursor, Orexin precursor, OrexinB, PPORX, PPOX | |
| Mouse | |
| Protein G purified | |
| Cancer, GPCR, Neuronal Cell Markers, Neuroscience | |
| 3060.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction