missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR5M1/OR5M10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16648764
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16648764 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16648764 Supplier Invitrogen™ Supplier No. PA5102290

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR5M1/5M10.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR5M1/OR5M10
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR5M10
Gene Accession No. Q6IEU7, Q8NGP8
Gene Alias olfactory receptor 5M1; Olfactory receptor 5M10; olfactory receptor family 5 subfamily M member 1; olfactory receptor family 5 subfamily M member 10; olfactory receptor OR11-207; Olfactory receptor OR11-208; olfactory receptor, family 5, subfamily M, member 1; olfactory receptor, family 5, subfamily M, member 10; OR11-207; OR11-208; OR5M1; OR5M10; OST050; seven transmembrane helix receptor
Gene Symbols OR5M1, OR5M10
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR5M1(Accession Q8NGP8), corresponding to amino acid residues F238-L288.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390167, 390168
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.