missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR10P1 Polyclonal Antibody

Product Code. 15809633
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15809633 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15809633 Supplier Invitrogen™ Supplier No. PA571220

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 85%; Guinea Pig: 92%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR10P1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OR10P1
Gene Accession No. Q8NGE3
Gene Alias Olfactory receptor 10P1; olfactory receptor 10P2; Olfactory receptor 10P3; olfactory receptor family 10 subfamily P member 1; olfactory receptor OR12-7; olfactory receptor, family 10, subfamily P, member 1 pseudogene; olfactory receptor, family 10, subfamily P, member 2 pseudogene; olfactory receptor, family 10, subfamily P, member 3 pseudogene; OR10P1; OR10P1P; OR10P2P; OR10P3P; OR12-7; OST701; seven transmembrane helix receptor
Gene Symbols OR10P1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human OR10P1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121130
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.