Learn More
SHP Steriltechnik AG Option M Laboklav 55-195, Recooling System
Description
- Highly effective recooling of liquids via a double jacket
- Cooling phase reduced by up to 50%, depending on bottle size and load quantity
- Full-surface chamber cooling instead of a coiled cooling coil
- Deionised water is reused via an integrated storage tank
- Heat is transferred via a heat exchanger to the chilled water system
- Significant savings in deionised water and operating costs
- Liquid loss during recooling approximately 5 to 8%
- Alternative options: Counter pressure option S or radial fan option L in cases of unavoidable liquid loss
Specifications
Specifications
| For Use With (Equipment) | Autoclaves, Laboklav 55-195 |
| Model | Option M Laboklav 55-195 |
| Product Type | Assisted Air Cooling |