Learn More
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Product Code. 10032851 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
10 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Pack of 50
This item is not returnable. Rückgaberichtlinie anzeigen
Artikelnummer. 10032851
missing translation for 'mfr': Macherey-Nagel™ 726824.25
Nur noch null auf Lager
Spezifikation
Diameter (Metric) 0.5 mm
Type High Performance GC Capillary Column
Diameter (Metric) Inner 0.32 mm
Stationary Phase Optima™ 1701
USP Type G46
For Use With (Application) Gas Chromatography (GC)
Temperature Range 300°C to 320°C
Film Thickness 0.35 μm
Length (Metric) 25 m
Quantity 1 Each