Description
- Open microplate bucket
- Compatible with Rotor S-4-104
- Available in packs of two and four
Specifications
Specifications
| For Use With (Equipment) | Rotor S-4-104 |
| Container Type | Microplate Bucket |
| Quantity | 4/Pk. |