Learn More
Melford ONPG
Product Code. 30018614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
Unit Size:
1g
25g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018614 50 g 50g
30018613 5 g 5g
30018611 1 g 1g
30018612 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30018614 Supplier Melford Supplier No. N8100050.0
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Colorimetric and spectrophotometric substrate for detection of β-D-Galactosidase.
Colorimetric and spectrophotometric substrate for detection of β-D-Galactosidase.
Specifications
Quantity 50 g